Anti-ABCA1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-12956R-CY3
10288-552EA 823.29 USD
10288-552
Anti-ABCA1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intracellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the ABC1 subfamily. Members of the ABC1 subfamily comprise the only major ABC subfamily found exclusively in multicellular eukaryotes. In humans, this protein functions as a cholesterol efflux pump in the cellular lipid removal pathway. Mutations in the human gene have been associated with Tangier's disease and familial high-density lipoprotein deficiency.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ABCA1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR