Anti-FCRLA Sheep Polyclonal Antibody

Supplier: R&D Systems
AF7965
102878-884EA 649.32 USD
102878-884
Anti-FCRLA Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen Symbol: FCRLA
Antigen Name: Fc receptor-like A
Antigen Synonyms: FCRL, FCRL1, FCRLM1, FCRLX, FCRLb, FCRLc1, FCRLc2, FCRLd, FCRLe, FCRX, FREB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR