Anti-PCK1 Sheep Polyclonal Antibody

Supplier: R&D Systems
AF7316
102878-600EA 649.32 USD
102878-600
Anti-PCK1 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PCK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR