Anti-FCRLA Sheep Polyclonal Antibody

Supplier: R&D Systems
AF7286
102878-578EA 649.32 USD
102878-578
Anti-FCRLA Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen Symbol: FCRL1
Antigen Name: Fc receptor-like 1
Antigen Synonyms: A230020G22Rik, Bxmas1, Bxmh1, Bxmh1b, Fcrh1, Fcrh1l, Fcrh1s, Ifgp1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR