Anti-ARX Sheep Polyclonal Antibody

Supplier: R&D Systems
AF7068
102878-214EA 750.55 USD
102878-214
Anti-ARX Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ARX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR