Anti-HPX Sheep Polyclonal Antibody

Supplier: R&D Systems
AF7007
102878-180EA 663.78 USD
102878-180
Anti-HPX Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HPX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR