Anti-VSIR Sheep Polyclonal Antibody

Supplier: R&D Systems
AF7005
102878-176EA 707.17 USD
102878-176
Anti-VSIR Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: VSIR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR