Anti-CIDEC Sheep Polyclonal Antibody

Supplier: R&D Systems
AF6960
102878-156EA 678.25 USD
102878-156
Anti-CIDEC Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CIDEC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR