WARMSTART® RTx Reverse Transcriptase, New England Biolabs
Supplier: New England Biolabs (NEB)
M0380L M0380S
102877-820EA 322 USD
102877-820 102877-822
WARMSTART® RTx Reverse Transcriptase, New England Biolabs
Nucleic Acid Reagents RT-PCR and cDNA Reagents
WarmStart RTx Reverse Transcriptase is a unique in silico designed RNA-directed DNA polymerase coupled with a reversibly-bound aptamer that inhibits RTx activity below 40°C
This enzyme can synthesize a complementary DNA strand initiating from a primer using RNA (cDNA synthesis) or single-stranded DNA as a template. RTx is a robust enzyme for RNA detection in amplification reactions and is particularly well suited for use in LAMP (loop-mediated isothermal amplification). The WarmStart property enables high throughput applications, room temperature setup, and increases the consistency and specificity of amplification reactions. RTx contains intact RNase H activity.