Clorox® Disinfecting Aerosol Spray
Supplier: Janitorial Supplies
CLO38504CT
102877-762CS 246.67 USD
102877-762
Clorox® Disinfecting Aerosol Spray
Cleaning Agents Disinfectants
Disinfecting aerosol spray.
  • Incredibly versatile and can be used on a wide variety of hard surfaces
  • Eliminates mold and kills flu virus
  • EPA-registered to kill as many as 64 microorganisms
  • Kills relevant bacteria, viruses and fungi that can cause serious illness and infection
  • Kills the majority of bacteria on its EPA Master Label in 3 minutes
  • Deodorizes and actually eliminates bacteria that cause bad odors
  • Ideal for use in offices, day care centers, schools, busy healthcare environments and other commercial facilities
Clorox® Disinfecting Spray kills relevant bacteria, viruses and fungi that can cause serious illness and infection. It is EPA-registered to kill as many as 64 microorganisms and it kills the majority of bacteria on its EPA Master Label in 3 minutes. With incredible versatility, Clorox® Disinfecting Spray can be used on a wide variety of hard surfaces while deodorizing and actually eliminating the bacteria that cause bad odors. From Clorox Commercial Solutions®, this disinfecting spray is ideal for use in offices, day care centers, schools, busy healthcare environments and other commercial facilities. Application: Disinfectant/Sanitizer; Applicable Material: Ceramics; Cement; Fiberglass; Glass; Grout; Laminates; Metal; Plastic; Vinyl; Dirt Types: Bacteria; Fungus; Germs; Mildew; Mold; Viruses; Scent: Fresh.