Anti-FOXD3 Goat Polyclonal Antibody

Supplier: R&D Systems
AF5090
102877-442EA 678.25 USD
102877-442
Anti-FOXD3 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen Symbol: FOXD3
Antigen Name: forkhead box D3
Antigen Synonyms: CWH3, Genesis, Hfh2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR