Anti-GLB1 Sheep Polyclonal Antibody

Supplier: R&D Systems
AF6464
102869-950EA 678.25 USD
102869-950
Anti-GLB1 Sheep Polyclonal Antibody
Antibodies
Simple Western

Type: Primary
Antigen Symbol: GLB1
Antigen Name: galactosidase, beta 1
Antigen Synonyms: EBP, ELNR1, MPS4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR