Anti-S100 A16 Sheep Polyclonal Antibody

Supplier: R&D Systems
AF6397
102869-902EA 678.25 USD
102869-902
Anti-S100 A16 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: S100 A16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR