Anti-SHISA4 Sheep Polyclonal Antibody

Supplier: R&D Systems
AF6365
102869-870EA 634.86 USD
102869-870
Anti-SHISA4 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SHISA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR