Anti-MUC1 Sheep Polyclonal Antibody

Supplier: R&D Systems
AF6298
102869-824EA 692.71 USD
102869-824
Anti-MUC1 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MUC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR