Anti-FGF10 Sheep Polyclonal Antibody

Supplier: R&D Systems
AF6224
102869-810EA 678.25 USD
102869-810
Anti-FGF10 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen Symbol: FGF10
Antigen Name: fibroblast growth factor 10
Antigen Synonyms: AEY17, BB213776, Fgf-10, Gsfaey17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR