Anti-CFHR4 Sheep Polyclonal Antibody

Supplier: R&D Systems
AF5980
102869-750EA 634.86 USD
102869-750
Anti-CFHR4 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CFHR4
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR