StabilCoat® Plus Stabilizer, Surmodics
Supplier: Surmodics
SC02-0050
102867-010EA 43.08 USD
102867-010
StabilCoat® Plus Stabilizer, Surmodics
Microarray Reagents
StabilCoat® Plus Stabilizer protects the activity of proteins while simultaneously blocking nonspecific binding sites on microspheres. In microarray applications, the conformation and activity of proteins are effectively preserved in a dried state when stored with StabilCoat® Plus stabilizer. The product ultimately increases signal and reduces background to improve the sensitivity of assays.
  • 3 year shelf life
  • Increased signal
  • Reduced background noise
  • Increased sensitivity
StabilCoat® Plus microarray stabilizer improves the stability of proteins covalently bound to microspheres stored in solution and array proteins stored in the dried state.
Certifications: ISO 13485:2016 & 9001:2015 Certified