BioFX® TMB Enhanced HRP Membrane Substrate
Supplier: Surmodics
The one component 3,3’,5,5’-tetramethylbenzidine (TMB) substrate is supplied in a ready-to-use solution for membrane applications. A highly intense staining pattern increases the signal-to-noise ratios for improved sensitivity in western blot, dot blots and line assays.
- Room temperature stability for 2 years
- Extended dynamic range with reduction of wash out at higher peroxidase concentrations
- Similar sensitivity to most chemiluminescent membrane substrates without the costly equipment needed for detection
BioFX® TMB Enhanced HRP Membrane Substrate provides an intense staining pattern for increased sensitivity.
Certifications: ISO 13485:2016 & 9001:2015 Certified