Spic and Span® Disinfecting All-Purpose Spray and Glass Cleaner
Supplier: Janitorial Supplies
PGC32538
102865-696CS 126.7 USD
102865-696
Spic and Span® Disinfecting All-Purpose Spray and Glass Cleaner
Cleaning Agents Disinfectants
This formula can replace a glass cleaner, an all-purpose cleaner, and a disinfecting cleaner and helps to save you both time and money.
  • Ammonia
  • Liquid
  • Unscented
For Ceramic; Fiberglass; Glass; Metal; Plastic; Vinyl. Dirt types include Bacteria; Dirt; Fungus; Germs; Grime; Mildew.
The patented, dual-solvent formula is designed to dissolve greasy soils and provide an easy wipe-up, then simply evaporate to leave your surface streak-free and disinfected. For those environments where disinfecting is of the utmost important, the effective formula is a hospital-grade disinfectant. This versatile cleaner can be used on most surfaces including glass.