Brecknell Electronic 60 lb. Coin and Parts Counting Scale, Essendant
Supplier: Janitorial Supplies
SBWB140
102865-602EA 377.51 USD
102865-602
Brecknell Electronic 60 lb. Coin and Parts Counting Scale, Essendant
Cash and Check Handling Supplies Coin Counters
Affordable, accurate 60 lbs. capacity parts or coin counting scale which is ideal for office or light industrial applications.
  • Configurable auto-off with rechargeable battery and RS-232
  • Units switch between lb or kg and also switch between currency
  • Rechargeable battery and AC adapter (both included)
  • Tare feature
Manufacturer's limited one-year warranty
Platform LxW: 11.5x8.75"
Increments: 1 g; 0.002 lbs.