E.coli RNA Polymerase, Core Enzyme, New England Biolabs
Supplier: New England Biolabs (NEB)
M0550S
102855-166EA
218
USD
102855-166
E.coli RNA Polymerase, Core Enzyme, New England Biolabs
Nucleic Acid Reagents
RNA Synthesis and Transcription
E.coli RNA Polymerase Core Enzyme consists of 5 subunits designated a, a, b´, β, and ω. The enzyme is free of sigma factor and does not initiate specific transcription from bacterial and phage DNA promoters. The enzyme retains the ability to transcribe RNA from nonspecific initiation sequences. Addition of sigma factors will allow the enzyme to initiate RNA synthesis from specific bacterial and phage promoters. The core enzyme has a molecular weight of approximately 400 kDa.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)