Rodent Insulin Chemiluminescence ELISA
Supplier: ALPCO DIAGNOSTICS MS
80-INSMR-CH01
102853-544EA 556.92 USD
102853-544
Rodent Insulin Chemiluminescence ELISA
Assays ELISAs
The Insulin Rodent (Mouse/Rat) Chemiluminescence ELISA is designed for the quantitative determination of insulin in rat and mouse serum, plasma, and tissue culture supernatants.
  • Flexibility to test mouse or rat samples
  • Low sample volume of 5 µl
  • Dynamic range of 0.1 to 150 ng/ml
  • Superior sensitivity of 0.089 ng/ml
Specifically, rat serum, rat heparin plasma, rat EDTA plasma, mouse serum, mouse heparin plasma, and tissue culture supernatants may be used in this assay.
The insulin rodent (mouse/rat) chemiluminescence ELISA is a sandwich type immunoassay. The 96-well microplate is coated with a monoclonal antibody specific for insulin. The standards, controls, and samples are added to the microplate wells with the conjugate. The microplate is then incubated at room temperature on a microplate shaker at 700 to 900 rpm. After the incubation is complete, the wells are washed with Wash Buffer and blotted dry. Chemiluminescent Substrate is added, and the microplate is read by a luminescence plate reader after 1 minute. The intensity of the light generated is directly proportional to the amount of insulin in the sample.
Caution: For research use only. Not for use in diagnostic procedures.