Porcine Albumin ELISA
Supplier: ALPCO DIAGNOSTICS MS
41-ALBPO-E01
102853-498EA 1120 USD
102853-498
Porcine Albumin ELISA
Assays ELISAs
The procine albumin ELISA is a highly sensitive two-site enzyme linked immunoassay for measuring albumin in porcine serum, tissue extracts and other biological fluids.
Albumin is a polyfunctional protein contributing to homeostasis through mechanisms of hemodynamics, transport and nutrition. Albumin is found both intra and extravascularly in all mammals and many lower vertebrates. It is a molecule of about 67,000 daltons, synthesized by the liver. Normally only very trace amounts of albumin escape reabsorption by kidney glomeruli and is excreted into the urine. Many occult diseases can cause kidney damage which may result in excessive amounts of serum proteins, including albumin, to be excreted by the kidney and into the urine.
Caution: For research use only. Not for use in diagnostic procedures.