Acetonitrile 25% (v/v) in water

Supplier: AQUA SOLUTIONS MS
Danger

M-521
102853-450EA 298.27 USD
102853-450
Acetonitrile 25% (v/v) in water
Acetonitrile
Formula: H₃CCN
MDL Number: MFCD00001878
CAS Number: 75-05-8
UN: 1648
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR