5-Acetyl-3-iodo-6-methyl-1,2-dihydropyridin-2-one ≥95%

Supplier: Matrix Scientific

087209-500MG 087209-1G
102852-908EA 803.08 USD
102852-908 102852-910
5-Acetyl-3-iodo-6-methyl-1,2-dihydropyridin-2-one ≥95%
5-Acetyl-3-iodo-6-methyl-1,2-dihydropyridin-2-one

Hazard: Irritant

Formula: C₈H₈INO₂
Melting Pt: 223…226 °C
MDL Number: MFCD22573522
CAS Number: 1407532-80-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR