2,2-Dimethyl-4'-methoxypropiophenone ≥97%

Supplier: Matrix Scientific

086756-1G 086756-2G 086756-5G
102850-438EA 672.31 USD
102850-438 102850-440 102850-442
2,2-Dimethyl-4'-methoxypropiophenone ≥97%
2,2-Dimethyl-4'-methoxypropiophenone

Hazard: Irritant

Formula: C₁₂H₁₆O₂
MDL Number: MFCD03841166
CAS Number: 2040-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR