2,2-Dimethyl-4'-methoxybutyrophenone ≥97%

Supplier: Matrix Scientific

086752-1G 086752-2G 086752-5G
102850-414EA 672.31 USD
102850-414 102850-416 102850-418
2,2-Dimethyl-4'-methoxybutyrophenone ≥97%
2,2-Dimethyl-4'-methoxybutyrophenone

Hazard: Irritant

Formula: C₁₃H₁₈O₂
MDL Number: MFCD03841121
CAS Number: 898765-08-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR