2-Phenyl-3'-(trifluormethyl)acetophenone ≥97%

Supplier: Matrix Scientific

086698-1G 086698-2G 086698-5G
102850-090EA 592.31 USD
102850-090 102850-092 102850-094
2-Phenyl-3'-(trifluormethyl)acetophenone ≥97%
2-Phenyl-3'-(trifluormethyl)acetophenone

Hazard: Irritant

Formula: C₁₅H₁₁F₃O
MDL Number: MFCD02260677
CAS Number: 1533-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR