8-Oxo-8-phenyloctanenitrile ≥97%
Supplier: Matrix Scientific
086670-1G
086670-2G
086670-5G
102849-922EA
347.69
USD
102849-922
102849-924
102849-926
8-Oxo-8-phenyloctanenitrile ≥97%
8-Oxo-8-phenyloctanenitrile
Hazard: Irritant
Formula:
C₁₄H₁₇NO |
MDL Number:
MFCD02260534 CAS Number: 39755-15-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...