8-Oxo-8-phenyloctanenitrile ≥97%

Supplier: Matrix Scientific

086670-1G 086670-2G 086670-5G
102849-922EA 347.69 USD
102849-922 102849-924 102849-926
8-Oxo-8-phenyloctanenitrile ≥97%
8-Oxo-8-phenyloctanenitrile

Hazard: Irritant

Formula: C₁₄H₁₇NO
MW: 215.3 g/mol
MDL Number: MFCD02260534
CAS Number: 39755-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR