4-(3-Bromobenzoyl)-2-methylpyridine ≥97%
Supplier: Matrix Scientific
086658-1G
086658-2G
086658-5G
102849-856EA
795.38
USD
102849-856
102849-858
102849-860
4-(3-Bromobenzoyl)-2-methylpyridine ≥97%
4-(3-Bromobenzoyl)-2-methylpyridine
Hazard: Irritant
Formula:
C₁₃H₁₀BrNO |
MDL Number:
MFCD13153289 CAS Number: 1187169-54-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...