2-(3-Methylbenzoyl)pyridine ≥97%

Supplier: Matrix Scientific

086599-1G 086599-2G 086599-5G
102849-508EA 795.38 USD
102849-508 102849-510 102849-512
2-(3-Methylbenzoyl)pyridine ≥97%
2-(3-Methylbenzoyl)pyridine

Hazard: Irritant

Formula: C₁₃H₁₁NO
MDL Number: MFCD07699226
CAS Number: 59576-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR