2-Chloro-3'-iodobenzophenone ≥97%

Supplier: Matrix Scientific

086535-1G 086535-2G 086535-5G
102849-124EA 387.69 USD
102849-124 102849-126 102849-128
2-Chloro-3'-iodobenzophenone ≥97%
2-Chloro-3'-iodobenzophenone

Hazard: Irritant

Formula: C₁₃H₈ClIO
MW: 342.57 g/mol
MDL Number: MFCD02260368
CAS Number: 890098-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR