3-(4-Methoxyphenyl)-2-methyl-1-propene ≥97%

Supplier: Matrix Scientific

086222-2G
102847-412EA 560 USD
102847-412
3-(4-Methoxyphenyl)-2-methyl-1-propene ≥97%
3-(4-Methoxyphenyl)-2-methyl-1-propene

Hazard: Irritant

Formula: C₁₁H₁₄O
MDL Number: MFCD07698654
CAS Number: 20849-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR