2-Nitro-1-naphthol ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Hydroxy-2-nitronaphthalene

085977-1G 085977-10G 085977-50G
102846-544EA 86.15 USD
102846-544 102846-546 102846-548
2-Nitro-1-naphthol ≥97%
2-Nitro-1-naphthol

Hazard: Irritant

Formula: C₁₀H₇NO₃
MW: 189.17
MDL Number: MFCD00003956
CAS Number: 607-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR