3,5,5-Trimethylcyclohexane-1,2-dione ≥97%

Supplier: Matrix Scientific

085856-1G 085856-10G 085856-50G
102846-208EA 72.31 USD
102846-208 102846-210 102846-212
3,5,5-Trimethylcyclohexane-1,2-dione ≥97%
3,5,5-Trimethylcyclohexane-1,2-dione

Hazard: Irritant

Formula: C₉H₁₄O₂
MDL Number: MFCD02181023
CAS Number: 57696-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR