Sucralose ≥97%
Supplier: Matrix Scientific
085854-1G
102846-204EA
383.08
USD
102846-204
Sucralose ≥97%
Sucralose
Sucralose
Purity: 97%
CAS: 56038-13-2
MDL Number: MFCD11100146
Molecular Formula: C12H19Cl3O8
Molecular Weight: 397,64
Hazard: Irritant
Formula:
C₁₂H₁₉Cl₃O₈ |
MDL Number:
MFCD03648615 CAS Number: 56038-13-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...