Cyclohexanecarbonitrile ≥97%

Supplier: Matrix Scientific

085805-1G 085805-10G
102846-032EA 30.77 USD
102846-032 102846-034
Cyclohexanecarbonitrile ≥97%
Cyclohexanecarbonitrile

Hazard: Irritant

Formula: C₇H₁₁N
MW: 109.17 g/mol
Boiling Pt: 75…76 °C (16 mmHg)
Melting Pt: 11 °C
Density: 0.919
Flash Pt: 65 °C
MDL Number: MFCD00040444
CAS Number: 766-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR