4-Chloro-6-(4-methylpiperazin-1-yl)-5-nitropyrimidine ≥97%
Supplier: Matrix Scientific
085455-1G
102845-088EA
666.15
USD
102845-088
4-Chloro-6-(4-methylpiperazin-1-yl)-5-nitropyrimidine ≥97%
4-Chloro-6-(4-methylpiperazin-1-yl)-5-nitropyrimidine
Hazard: Irritant
Formula:
C₉H₁₂ClN₅O₂ |
MDL Number:
MFCD02945691 CAS Number: 154418-73-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...