4-tert-Butyl-2-methylthiazole ≥97%
Supplier: Matrix Scientific
085389-1G
085389-10G
102844-918EA
127.69
USD
102844-918
102844-920
4-tert-Butyl-2-methylthiazole ≥97%
4-tert-Butyl-2-methylthiazole
Purity: 97%
CAS: 15679-11-5
MDL Number: MFCD00051643
Molecular Formula: C8H13NS
Molecular Weight: 155,26
Hazard: Irritant
Formula:
C₈H₁₃NS |
MDL Number:
MFCD00051643 CAS Number: 15679-11-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...