1-Ethyl-3-nitro-1H-pyrazole ≥97%

Supplier: Matrix Scientific

085383-1G
102844-904EA 723.08 USD
102844-904
1-Ethyl-3-nitro-1H-pyrazole ≥97%
1-Ethyl-3-nitro-1H-pyrazole

Hazard: Irritant

Formula: C₅H₇N₃O₂
MDL Number: MFCD04970018
CAS Number: 58793-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR