4-Propylphenol ≥97%
Supplier: Matrix Scientific
Synonyms:
p-Propylphenol, 4-n-Propylphenol
085010-1G
085010-10G
102844-018EA
143.08
USD
102844-018
102844-020
4-Propylphenol ≥97%
4-Propylphenol
Hazard: Irritant
Formula:
C₉H₁₂O |
MDL Number:
MFCD00002395 CAS Number: 645-56-7 UN: 3145 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...