1-Isopropenyl-4-methoxybenzene ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Methoxy-4-(1-propen-2-yl)benzene, 4-Isopropenylanisole

085009-1G
102844-016EA 806.15 USD
102844-016
1-Isopropenyl-4-methoxybenzene ≥97%
1-Isopropenyl-4-methoxybenzene

Hazard: Irritant

Formula: C₁₀H₁₂O
MW: 148.21 g/mol
MDL Number: MFCD09414742
CAS Number: 1712-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR