1-(3,4-Dichlorophenyl)ethan-1-one oxime ≥97%

Supplier: Matrix Scientific

084971-1G 084971-10G
102843-872EA 213.85 USD
102843-872 102843-874
1-(3,4-Dichlorophenyl)ethan-1-one oxime ≥97%
1-(3,4-Dichlorophenyl)ethan-1-one oxime

Hazard: Irritant

Formula: C₈H₇Cl₂NO
MDL Number: MFCD00084991
CAS Number: 71516-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR