N4-Ethyl-N4-(2-hydroxyethyl)-2-methyl-1,4-phenylenediamine sulfate ≥97%
Supplier: Matrix Scientific
084913-10G
084913-50G
102843-718EA
116.92
USD
102843-718
102843-720
N4-Ethyl-N4-(2-hydroxyethyl)-2-methyl-1,4-phenylenediamine sulfate ≥97%
N4-Ethyl-N4-(2-hydroxyethyl)-2-methyl-1,4-phenylenediamine sulfate
Purity: 97%
CAS: 25646-77-9
MDL Number: MFCD00035563
Molecular Formula: C11H20N2O5S
Molecular Weight: 292,36
Hazard: Irritant
Formula:
C₁₁H₁₈N₂O |
CAS Number:
25646-77-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...