5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB) ≥97%

Supplier: Matrix Scientific

Synonyms: DTNB, Ellman's reagent, 5,5'-Disulfanediylbis(2-nitrobenzoic acid), 3,3'-Dithiobis(6-nitrobenzoic acid), 5,5'-Disulphanediylbis(2-nitrobenzoic acid), 2,2'-Dinitro-5,5'-dithiodibenzoic acid

084824-50G
102843-490EA 849.23 USD
102843-490
5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB) ≥97%
5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB)

Hazard: Irritant

Formula: C₁₄H₈N₂O₈S₂
MW: 396.36 g/mol
MDL Number: MFCD00007140
CAS Number: 69-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR