3-(Trifluoromethyl)phenylacetone ≥97%

Supplier: Matrix Scientific

Synonyms: 1-[3-(Trifluoromethyl)phenyl]-2-propanone

084687-50G
102843-094EA 481.54 USD
102843-094
3-(Trifluoromethyl)phenylacetone ≥97%
3-(Trifluoromethyl)phenylacetone

Hazard: Irritant

Formula: C₁₀H₉F₃O
MDL Number: MFCD00000397
CAS Number: 21906-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR