1-(2-Methoxyphenyl)-2-thiourea ≥97%

Supplier: Matrix Scientific

Synonyms: N-(2-Methoxyphenyl)thiourea, 1-(2-Methoxyphenyl)thiourea, 2-Methoxyphenylthiourea

084644-1G 084644-10G 084644-50G
102842-976EA 101.54 USD
102842-976 102842-978 102842-980
1-(2-Methoxyphenyl)-2-thiourea ≥97%
1-(2-Methoxyphenyl)-2-thiourea

Hazard: Irritant

Formula: (C₆H₄(OCH₃))NC(S)NH₂
MDL Number: MFCD00022162
CAS Number: 1516-37-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR