1-(2-Methoxyphenyl)-2-thiourea ≥97%
Supplier: Matrix Scientific
Synonyms:
N-(2-Methoxyphenyl)thiourea, 1-(2-Methoxyphenyl)thiourea, 2-Methoxyphenylthiourea
084644-1G
084644-10G
084644-50G
102842-976EA
101.54
USD
102842-976
102842-978
102842-980
1-(2-Methoxyphenyl)-2-thiourea ≥97%
1-(2-Methoxyphenyl)-2-thiourea
Hazard: Irritant
Formula:
(C₆H₄(OCH₃))NC(S)NH₂ |
MDL Number:
MFCD00022162 CAS Number: 1516-37-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...